The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145445.3 | NP_663420.1 | 1 - 2 | 210 | 72 |
GenBank mRNA | AK049731.1 | - | 1 - 2 | 211 | 72 |
AK076195.1 | - | 1 - 2 | 206 | 72 | |
BC003326.1 | - | 1 - 2 | 242 | 72 |