Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syndig1l
Entrez Gene ID
627191
Gene Name
synapse differentiation inducing 1 like
Gene Aliases
DSPC1, Gm261, Tmem90a
Chromosome Location
Chr.12: 84677277 - 84698907 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033334.2 NP_001028506.1 1 - 2 281 136
GenBank mRNA AK140195.1 - 1 - 2 281 136
AY993934.1 - 1 - 2 305 136

View Full Product Details