Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Abcd4
Entrez Gene ID
19300
Gene Name
ATP-binding cassette, sub-family D (ALD), member 4
Gene Aliases
P69r, P70R, Pxmp1l
Chromosome Location
Chr.12: 84602531 - 84617479 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008992.2 NP_033018.2 18 - 19 1899 69
GenBank mRNA AJ001166.1 - 18 - 19 1803 69
AK028420.1 - 18 - 19 1956 69

View Full Product Details