The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175382.3 | NP_780591.1 | 1 - 2 | 870 | 105 |
GenBank mRNA | AK041641.1 | - | 1 - 2 | 872 | 105 |
AK076058.1 | - | 1 - 2 | 892 | 105 | |
BC090647.1 | - | 1 - 2 | 956 | 105 | |
BC106123.1 | - | 1 - 2 | 854 | 105 | |
BC138225.1 | - | 1 - 2 | 839 | 105 | |
BC145667.1 | - | 1 - 2 | 839 | 105 |