Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Rffl
- Entrez Gene ID
-
67338
- Gene Name
- ring finger and FYVE like domain containing protein
- Gene Aliases
-
1700051E09Rik, 4930516L10Rik, BG080975, Carp2
- Chromosome Location
-
Chr.11: 82803819 - 82871239 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK015806.1 |
- |
4 - 5 |
1125 |
65 |
View Full Product Details