Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Akap1
Entrez Gene ID
11640
Gene Name
A kinase (PRKA) anchor protein 1
Gene Aliases
AKAP121, AKAP84, Akap, C76494, C81186, DAKAP1, S-AKAP84
Chromosome Location
Chr.11: 88830792 - 88864586 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042541.1 NP_001036006.1 7 - 8 2512 88
NM_009648.2 NP_033778.2 7 - 8 2475 88
NR_104310.1 8 - 9 2591 88
GenBank mRNA AK076216.1 - 7 - 8 2515 88
AK132749.1 - 7 - 8 2486 88
BC065135.1 - 7 - 8 2424 88
U95145.1 - 8 - 9 2595 88
U95146.1 - 7 - 8 2488 88

View Full Product Details