The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_036613.1 | 3 - 4 | 469 | 80 | |
GenBank mRNA | AK152411.1 | - | 3 - 4 | 457 | 80 |
AK152779.1 | - | 3 - 4 | 457 | 80 | |
AK153073.1 | - | 3 - 4 | 457 | 80 |