The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039181.1 | NP_001034270.1 | 7 - 8 | 1529 | 69 |
NM_001286395.1 | NP_001273324.1 | 8 - 9 | 1145 | 69 | |
NM_008728.2 | NP_032754.2 | 7 - 8 | 1532 | 69 | |
GenBank mRNA | AA763613.1 | - | 2 - 3 | 212 | 69 |
AK079196.1 | - | 8 - 9 | 1062 | 69 | |
AK166535.1 | - | 6 - 7 | 760 | 69 | |
BC055897.1 | - | 7 - 8 | 1529 | 69 | |
D78175.1 | - | 7 - 8 | 1497 | 69 |