The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175251.4 | NP_780460.3 | 20 - 21 | 5512 | 102 |
GenBank mRNA | AK014481.1 | - | 7 - 8 | 3477 | 102 |
AK034964.1 | - | 6 - 7 | 1673 | 102 | |
AK036304.1 | - | 1 - 2 | 91 | 102 | |
AK077022.1 | - | 4 - 5 | 1751 | 102 | |
AK077366.1 | - | 6 - 7 | 2274 | 102 | |
AK173201.1 | - | 6 - 7 | 3339 | 102 | |
BC027180.1 | - | 6 - 7 | 2411 | 102 |