The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009546.2 | NP_033572.2 | 1 - 2 | 677 | 77 |
GenBank mRNA | AK154533.1 | - | 1 - 2 | 670 | 77 |
AK160898.1 | - | 1 - 2 | 629 | 77 | |
AK169562.1 | - | 1 - 2 | 678 | 77 | |
AK171744.1 | - | 1 - 2 | 321 | 77 | |
BC006908.1 | - | 1 - 2 | 646 | 77 | |
BC034276.1 | - | 1 - 2 | 649 | 77 | |
D63902.1 | - | 1 - 2 | 600 | 77 |