The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195003.1 | NP_001181932.1 | 1 - 2 | 213 | 95 |
NM_001195004.1 | NP_001181933.1 | 1 - 2 | 213 | 95 | |
NM_177619.3 | NP_808287.1 | 1 - 2 | 213 | 95 | |
GenBank mRNA | AK169458.1 | - | 1 - 2 | 135 | 95 |
AK170389.1 | - | 1 - 2 | 160 | 95 | |
BC048904.1 | - | 1 - 2 | 168 | 95 | |
BC057102.1 | - | 1 - 2 | 141 | 95 | |
DQ076247.1 | - | 1 - 2 | 141 | 95 |