Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcnx
Entrez Gene ID
54604
Gene Name
pecanex homolog (Drosophila)
Gene Aliases
2900024E21Rik, 3526401J03Rik, AF096286, AI327143, AI413187, Pcnxl1
Chromosome Location
Chr.12: 81860028 - 82000924 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018814.3 NP_061284.2 11 - 12 3350 96
GenBank mRNA AF096286.1 - 5 - 6 1180 96
AK129221.1 - 6 - 7 1230 96
AK135724.1 - 5 - 6 806 96
BC057333.1 - 4 - 5 1093 96
BC066849.1 - 6 - 7 1023 96
BC098370.1 - 6 - 7 940 96

View Full Product Details