Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcnx
Entrez Gene ID
54604
Gene Name
pecanex homolog (Drosophila)
Gene Aliases
2900024E21Rik, 3526401J03Rik, AF096286, AI327143, AI413187, Pcnxl1
Chromosome Location
Chr.12: 81860028 - 82000924 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018814.3 NP_061284.2 15 - 16 3820 117
GenBank mRNA AF096286.1 - 9 - 10 1650 117
AK129221.1 - 10 - 11 1700 117
BC057333.1 - 8 - 9 1563 117
BC098370.1 - 10 - 11 1410 117

View Full Product Details