Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rpl23
Entrez Gene ID
65019
Gene Name
ribosomal protein L23
Gene Aliases
2810009A01Rik
Chromosome Location
Chr.11: 97777526 - 97782439 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK139855.1 - 1 - 2 259 89

View Full Product Details