Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tbc1d24
Entrez Gene ID
224617
Gene Name
TBC1 domain family, member 24
Gene Aliases
9630033P11, C530046L02Rik, mKIAA1171
Chromosome Location
Chr.17: 24175431 - 24205562 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163848.1 NP_001157320.1 9 - 10 2020 59
NM_001163849.1 NP_001157321.1 9 - 10 1934 59
NM_001163851.1 NP_001157323.1 8 - 9 2002 59
GenBank mRNA AK083833.1 - 9 - 10 1906 59
AK086293.1 - 8 - 9 1961 59
AK138431.1 - 9 - 10 1895 59
AK147647.1 - 9 - 10 2020 59

View Full Product Details