Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Tspan2os
- Entrez Gene ID
-
78491
- Gene Name
- tetraspanin 2, opposite strand
- Gene Aliases
-
2410057H14Rik
- Chromosome Location
-
Chr.3: 102720231 - 102735417 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_040588.1 |
|
2 - 3 |
280 |
122 |
GenBank mRNA |
AK019118.1 |
- |
2 - 3 |
282 |
122 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BY710128.1 |
- |
2 - 3 |
282 |
|
Chr. 3 - 102720231 - 102735419 [-] on Build GRCm38 |
122 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
nerve growth factor |
Ngf |
Ngfb |
-205.3kb
|
Anti-sense |
Chr. - 102469919 - 102521013 on Build GRCm38 |
Downstream |
tetraspanin 2 |
Tspan2 |
6330415F13Rik, B230119D02Rik, tspan-2 |
7.7kb
|
Anti-sense |
Chr. - 102734770 - 102772310 on Build GRCm38 |
View Full Product Details