The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007497.3 | NP_031523.3 | 6 - 7 | 901 | 104 |
GenBank mRNA | AK144511.1 | - | 6 - 7 | 944 | 104 |
BC006871.1 | - | 6 - 7 | 847 | 104 | |
M63725.1 | - | 5 - 6 | 691 | 104 |