Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Syngap1
Entrez Gene ID
240057
Gene Name
synaptic Ras GTPase activating protein 1 homolog (rat)
Gene Aliases
Gm1963, Syngap
Chromosome Location
Chr.17: 26941408 - 26972434 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281491.1 NP_001268420.1 11 - 12 1920 76
GenBank mRNA CB525601.1 - 2 - 3 358 76

View Full Product Details