Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syngap1
Entrez Gene ID
240057
Gene Name
synaptic Ras GTPase activating protein 1 homolog (rat)
Gene Aliases
Gm1963, Syngap
Chromosome Location
Chr.17: 26941408 - 26972434 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281491.1 NP_001268420.1 2 - 3 184 59

View Full Product Details