The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001207021.1 | NP_001193950.1 | 3 - 4 | 1207 | 82 |
GenBank mRNA | AW213062.1 | - | 3 - 4 | 373 | 82 |
BG919271.1 | - | - | 291 | 82 | |
BU558590.1 | - | 3 - 4 | 414 | 82 |