The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009196.4 | NP_033222.1 | 4 - 5 | 1414 | 80 |
GenBank mRNA | AF058055.1 | - | 4 - 5 | 1395 | 80 |
AK033516.1 | - | 4 - 5 | 1412 | 80 | |
AK053421.1 | - | 1 - 2 | 775 | 80 | |
AK079379.1 | - | 4 - 5 | 1410 | 80 | |
AK088762.1 | - | 4 - 5 | 1412 | 80 | |
AK136198.1 | - | 1 - 2 | 407 | 80 | |
AK161701.1 | - | 4 - 5 | 1412 | 80 | |
AK166606.1 | - | 4 - 5 | 1412 | 80 | |
BC014777.1 | - | 4 - 5 | 1422 | 80 | |
X82438.1 | - | 3 - 4 | 1206 | 80 |