The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207533.2 | NP_997416.2 | 1 - 2 | 535 | 59 |
GenBank mRNA | AK035803.1 | - | 1 - 2 | 132 | 59 |
CX214957.1 | - | 1 - 2 | 452 | 59 |