Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hiatl1
Entrez Gene ID
66631
Gene Name
hippocampus abundant transcript-like 1
Gene Aliases
5730414C17Rik, AU045608, AW539692
Chromosome Location
Chr.13: 65065030 - 65113000 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083901.1 NP_001077370.1 3 - 4 472 100
NM_133680.3 NP_598441.3 2 - 3 569 100
GenBank mRNA AK146596.1 - 3 - 4 465 100
BC033469.1 - 2 - 3 531 100

View Full Product Details