The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025661.4 | NP_079937.1 | 1 - 2 | 118 | 81 |
GenBank mRNA | AK087497.1 | - | 1 - 2 | 87 | 81 |
AK154085.1 | - | 1 - 2 | 110 | 81 | |
BC046594.1 | - | 1 - 2 | 56 | 81 |