Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tex14
Entrez Gene ID
83560
Gene Name
testis expressed gene 14
Gene Aliases
C85585
Chromosome Location
Chr.11: 87405050 - 87555823 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199293.1 NP_001186222.1 30 - 31 4440 63
NM_031386.2 NP_113563.2 30 - 31 4421 63
GenBank mRNA AK036355.1 - 4 - 5 476 63


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW554038.1 - Chr. - - [-] on Build 63

View Full Product Details