The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183017.2 | NP_898838.2 | 13 - 14 | 1809 | 92 |
GenBank mRNA | AK084879.1 | - | 13 - 14 | 1777 | 92 |
AK143496.1 | - | 12 - 13 | 1642 | 92 | |
AK150116.1 | - | 13 - 14 | 1775 | 92 | |
AM690756.1 | - | 13 - 14 | 1762 | 92 | |
BC055368.1 | - | 13 - 14 | 1774 | 92 |