Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Dnal1
- Entrez Gene ID
-
105000
- Gene Name
- dynein, axonemal, light chain 1
- Gene Aliases
-
1700010H15Rik, AW121714, Dnalc1, E330027P08Rik
- Chromosome Location
-
Chr.12: 84114280 - 84143517 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK005826.1 |
- |
1 - 2 |
28 |
75 |
View Full Product Details