The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008702.2 | NP_001008702.1 | 4 - 5 | 535 | 101 |
NM_001037905.3 | NP_001032994.1 | 4 - 5 | 609 | 101 | |
NM_001102400.1 | NP_001095870.1 | 4 - 5 | 609 | 101 | |
NM_023118.5 | NP_075607.2 | 4 - 5 | 609 | 101 | |
GenBank mRNA | AK002850.1 | - | 4 - 5 | 492 | 101 |
AK154716.1 | - | 4 - 5 | 593 | 101 | |
AK162744.1 | - | 4 - 5 | 580 | 101 | |
AK162851.1 | - | 4 - 5 | 580 | 101 | |
AK165519.1 | - | 4 - 5 | 493 | 101 | |
AK165810.1 | - | 4 - 5 | 490 | 101 | |
AK168375.1 | - | 4 - 5 | 581 | 101 | |
BC006588.1 | - | 3 - 4 | 430 | 101 | |
BC016887.1 | - | 4 - 5 | 475 | 101 | |
CD348957.1 | - | 2 - 3 | 132 | 101 | |
U18869.1 | - | 4 - 5 | 542 | 101 |