The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011019.3 | NP_035149.2 | 17 - 18 | 2481 | 84 |
GenBank mRNA | AB015978.1 | - | 17 - 18 | 3132 | 84 |
AF058805.1 | - | 17 - 18 | 2451 | 84 | |
BC137703.1 | - | 16 - 17 | 2379 | 84 |