The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008533.3 | NP_001008533.1 | 1 - 2 | 1442 | 58 |
NM_001039510.2 | NP_001034599.1 | 2 - 3 | 627 | 58 | |
NM_001282945.1 | NP_001269874.1 | 2 - 3 | 661 | 58 | |
GenBank mRNA | AJ555877.1 | - | 1 - 2 | 340 | 58 |
AK038774.1 | - | 2 - 3 | 601 | 58 | |
AK043954.1 | - | 1 - 2 | 1383 | 58 | |
AK045729.1 | - | 1 - 2 | 982 | 58 | |
AK134532.1 | - | 2 - 3 | 630 | 58 | |
AK134553.1 | - | 1 - 2 | 847 | 58 | |
AK134709.1 | - | 1 - 2 | 808 | 58 | |
AU080986.1 | - | 2 - 3 | 568 | 58 | |
BC026602.1 | - | 1 - 2 | 293 | 58 | |
BC079624.1 | - | 1 - 4 | 563 | 58 | |
U05671.1 | - | 1 - 2 | 647 | 58 |