Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Chkb
- Entrez Gene ID
-
12651
- Gene Name
- choline kinase beta
- Gene Aliases
-
Chetk, Chkl, Ck, Ck/Ek, Ck/Ek-beta, Ckb, Ckekb, Ek
- Chromosome Location
-
Chr.15: 89426349 - 89429927 on Build GRCm38
- Gene Symbol
- ChkbCpt1b
- Entrez Gene ID
-
100134861
- Gene Name
- Chkb-Cpt1b readthrough transcript (NMD candidate)
- Gene Aliases
-
BC090627, Chkb-Cpt1b, Chkbcpt1b
- Chromosome Location
-
Chr.15: 89416405 - 89429927 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_004843.2 |
|
9 - 10 |
1612 |
82 |
View Full Product Details