The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027306.3 | NP_081582.1 | 1 - 1 | 1031 | 96 |
GenBank mRNA | AK131706.1 | - | 1 - 1 | 1005 | 96 |
BC049767.1 | - | 1 - 1 | 1031 | 96 |