The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033303.2 | NP_001028475.2 | 15 - 16 | 2154 | 65 |
GenBank mRNA | AK132524.1 | - | 15 - 16 | 2145 | 65 |