Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zscan10
Entrez Gene ID
332221
Gene Name
zinc finger and SCAN domain containing 10
Gene Aliases
BC068284, Zfp206, Zkscan10, Znf206
Chromosome Location
Chr.17: 23599751 - 23611019 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033425.4 NP_001028597.2 2 - 3 588 62
NM_001289481.1 NP_001276410.1 2 - 3 588 62
GenBank mRNA AK141259.1 - 2 - 3 559 62
BC145638.1 - 1 - 2 421 62
CF896527.1 - 2 - 3 489 62
DQ323929.1 - 2 - 3 556 62
DQ323930.1 - 2 - 3 556 62
EF152498.1 - 1 - 2 403 62

View Full Product Details