Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Phtf1
Entrez Gene ID
18685
Gene Name
putative homeodomain transcription factor 1
Gene Aliases
AU041898, AW049785, Phft, Phtf
Chromosome Location
Chr.3: 103968110 - 104007490 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163467.1 NP_001156939.1 9 - 10 1117 106
NM_001163468.1 NP_001156940.1 8 - 9 1364 106
NM_001163469.1 NP_001156941.1 8 - 9 1388 106
NM_013629.2 NP_038657.2 9 - 10 1523 106
GenBank mRNA AJ133721.1 - 9 - 10 1117 106
AJ242864.1 - 9 - 10 1523 106
AK019146.1 - 9 - 10 1427 106
AK088714.1 - 8 - 9 1255 106
AK131629.1 - - 1414 106
BC021147.1 - 9 - 10 1396 106
BC050825.1 - 8 - 9 1200 106
BC057340.1 - 9 - 10 1409 106
BC060177.1 - 9 - 10 1409 106

View Full Product Details