The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301304.1 | NP_001288233.1 | 5 - 6 | 1582 | 152 |
NM_001301306.1 | NP_001288235.1 | 5 - 6 | 1692 | 152 | |
NM_010432.2 | NP_034562.2 | 5 - 6 | 1692 | 152 | |
NR_125731.1 | 5 - 6 | 1580 | 152 | ||
GenBank mRNA | AF071070.1 | - | 5 - 6 | 1582 | 152 |
AF071071.1 | - | 5 - 6 | 1580 | 152 | |
AF077658.1 | - | 5 - 6 | 1596 | 152 | |
AF170303.1 | - | 5 - 6 | 1492 | 152 | |
AF170304.1 | - | 5 - 6 | 1492 | 152 | |
AK134408.1 | - | 5 - 6 | 152 | ||
BC145697.1 | - | 5 - 6 | 1445 | 152 | |
BC145699.1 | - | 5 - 6 | 1445 | 152 | |
DQ279085.1 | - | 3 - 4 | 263 | 152 | |
DQ279089.1 | - | 3 - 4 | 239 | 152 |