Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mks1
Entrez Gene ID
380718
Gene Name
Meckel syndrome, type 1
Gene Aliases
AK190930, B8d3
Chromosome Location
Chr.11: 87852928 - 87863679 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039684.2 NP_001034773.2 2 - 3 215 79
GenBank mRNA AK030982.1 - 2 - 3 195 79
AK138267.1 - 1 - 2 185 79
AK170482.1 - 2 - 3 213 79
BC150995.1 - 2 - 3 283 79
BC158122.1 - 2 - 3 375 79
DQ177342.1 - 2 - 3 215 79

View Full Product Details