Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Flywch2
Entrez Gene ID
76917
Gene Name
FLYWCH family member 2
Gene Aliases
2810417J12Rik, 3010014L17Rik, AW548203
Chromosome Location
Chr.17: 23776918 - 23786077 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029798.3 NP_084074.1 1 - 2 81 69
GenBank mRNA AK013108.1 - 1 - 2 83 69

View Full Product Details