The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033339.3 | NP_001028511.1 | 6 - 7 | 1502 | 107 |
GenBank mRNA | AK089273.1 | - | 6 - 7 | 1241 | 107 |
AK089533.1 | - | 6 - 7 | 1241 | 107 | |
AK137491.1 | - | 6 - 7 | 1104 | 107 | |
AK154458.1 | - | 6 - 7 | 1504 | 107 | |
AK171493.1 | - | 6 - 7 | 1241 | 107 | |
BC112379.1 | - | 6 - 7 | 1188 | 107 |