The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023058.3 | NP_075545.2 | 2 - 3 | 795 | 64 |
GenBank mRNA | AF175892.1 | - | 2 - 3 | 679 | 64 |
AK158550.1 | - | 2 - 3 | 796 | 64 | |
AK165164.1 | - | 2 - 3 | 662 | 64 | |
AK181294.1 | - | 1 - 2 | 37 | 64 | |
BC025061.1 | - | 2 - 3 | 518 | 64 |