Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adam21
Entrez Gene ID
56622
Gene Name
a disintegrin and metallopeptidase domain 21
Gene Aliases
Adam31
Chromosome Location
Chr.12: 81558584 - 81568474 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020330.4 NP_065063.1 1 - 2 389 65
GenBank mRNA AK014827.2 - 1 - 2 390 65

View Full Product Details