Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcnx
Entrez Gene ID
54604
Gene Name
pecanex homolog (Drosophila)
Gene Aliases
2900024E21Rik, 3526401J03Rik, AF096286, AI327143, AI413187, Pcnxl1
Chromosome Location
Chr.12: 81860028 - 82000924 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018814.3 NP_061284.2 8 - 9 2974 69
GenBank mRNA AK129221.1 - 3 - 4 854 69
BC098370.1 - 3 - 4 579 69

View Full Product Details