The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009401.2 | NP_033427.1 | 12 - 13 | 1314 | 62 |
GenBank mRNA | BC130026.1 | - | 12 - 13 | 1305 | 62 |
U25416.1 | - | 12 - 13 | 1314 | 62 |