The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114334.1 | NP_001107806.1 | 1 - 2 | 239 | 100 |
NM_028259.4 | NP_082535.1 | 1 - 2 | 239 | 100 | |
GenBank mRNA | AK032724.1 | - | 1 - 2 | 201 | 100 |
AK085687.1 | - | 1 - 2 | 208 | 100 | |
AK163258.1 | - | 1 - 2 | 209 | 100 | |
AK178169.1 | - | 1 - 2 | 121 | 100 | |
BC038491.1 | - | 1 - 2 | 212 | 100 | |
BC071244.1 | - | 1 - 2 | 196 | 100 |