Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gtf2f2
Entrez Gene ID
68705
Gene Name
general transcription factor IIF, polypeptide 2
Gene Aliases
1110031C13Rik, AI326315
Chromosome Location
Chr.14: 75896937 - 76010865 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026816.3 NP_081092.1 6 - 7 599 102
GenBank mRNA AK144858.1 - 6 - 7 600 102
BC027173.1 - 6 - 7 595 102

View Full Product Details