Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd13c
Entrez Gene ID
433667
Gene Name
ankyrin repeat domain 13c
Gene Aliases
AI505652, AU022220
Chromosome Location
Chr.3: 157946668 - 158006837 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013806.1 NP_001013828.1 10 - 11 1360 72
GenBank mRNA AK135906.1 - 11 - 12 1096 72
AK157542.1 - - 1542 72
BC065088.1 - 9 - 10 3554 72
BC138991.1 - 10 - 11 1368 72
BC138992.1 - 11 - 12 1347 72
CF736962.1 - 3 - 4 356 72

View Full Product Details