The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146193.2 | NP_666305.2 | 1 - 2 | 455 | 97 |
GenBank mRNA | AK088345.1 | - | 1 - 2 | 496 | 97 |
BC072618.1 | - | 1 - 2 | 455 | 97 | |
BC087937.1 | - | 1 - 2 | 488 | 97 |