Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc81
Entrez Gene ID
70884
Gene Name
coiled-coil domain containing 81
Gene Aliases
4921513D09Rik
Chromosome Location
Chr.7: 89866148 - 89903629 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162979.1 NP_001156451.1 1 - 2 445 79
GenBank mRNA AK014876.1 - 1 - 2 428 79
AK029544.1 - 1 - 2 446 79
BC127049.1 - 1 - 2 432 79
BC152307.1 - 1 - 2 434 79
BC152308.1 - 1 - 2 434 79

View Full Product Details