The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277957.1 | NP_001264886.1 | 1 - 2 | 213 | 67 |
NM_021466.3 | NP_067441.1 | 1 - 2 | 213 | 67 | |
GenBank mRNA | AK040747.1 | - | 1 - 2 | 117 | 67 |
BC061106.1 | - | 1 - 2 | 98 | 67 | |
BC138425.1 | - | 1 - 2 | 79 | 67 | |
CN707961.1 | - | 1 - 2 | 213 | 67 | |
Y09972.1 | - | 1 - 2 | 115 | 67 |