Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pex1
Entrez Gene ID
71382
Gene Name
peroxisomal biogenesis factor 1
Gene Aliases
5430414H02Rik, E330005K07Rik, ZWS1
Chromosome Location
Chr.5: 3596066 - 3637230 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293806.1 NP_001280735.1 21 - 22 3767 70
NM_027777.2 NP_082053.1 20 - 21 3647 70
GenBank mRNA AK017309.1 - 8 - 9 1064 70
BC090845.1 - 20 - 21 3647 70

View Full Product Details